Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KV3.1 (KCNC1) Rabbit anti-Bovine, Canine, Human, Mouse, Rabbit, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523012

Catalog No. PIPA523012


Add to cart

Description

Description

Suggested positive control: HEK293 cell lysate.

Voltage-gated K+ channels are important determits of neuronal membrane excitability. Moreover, differences in K+ channel expression patterns and densities contribute to the variations in action potential waveforms and repetitive firing patterns evident in different neuronal cell types (Maletic-Savatic et al., 1995; Pongs, 1999; Blaine and Ribera, 1998; Burger and Ribera, 1996). The Kv3.1 potassium channel is expressed at high levels in neurons that characteristically fire rapid trains of action potentials (Gan et al., 1999). Particularly high levels of this channel are found in neurons of the auditory brainstem. These neurons appear to participate in neural circuits that determine the intensity and timing of auditory stimuli and use this information to determine the location of sounds in space (von Hehn et al., 2004).
Specifications

Specifications

KV3.1 (KCNC1)
Polyclonal
Unconjugated
KCNC1
Liquid
KCNC1
Synthetic peptide made to an internal portion of rat Kv3.1 (within residues 350-400).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Canine, Human, Mouse, Rabbit, Rat
Immunoprecipitation, Neutralization, Western Blot
1 mg/mL
PBS with 0.1% sodium azide
P48547, P15388, P25122
KV3.1, KV4, NGK2, potassium voltage-gated channel subfamily C member 1, voltage-gated potassium channel protein KV3.1, voltage-gated potassium channel subunit Kv3.1, voltage-gated potassium channel subunit Kv4
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100008968, 16502, 25327, 3746, 403705, 541459
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit