Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Kv7.5, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136968

Catalog No. NBP136968


Add to cart

Description

Description

Kv7.5 Polyclonal antibody specifically detects Kv7.5 in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Peptide ELISA.
Specifications

Specifications

Kv7.5
0.5 mg/mL
Peptide ELISA Detection limit 1:64000
NP_062816.2
KQT-like 5, Kv7.5, potassium channel protein, Potassium channel subunit alpha KvLQT5, potassium voltage-gated channel subfamily KQT member 5, potassium voltage-gated channel, KQT-like subfamily, member 5, Voltage-gated potassium channel subunit Kv7.5
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to cross-react with isoform 1, 2, 3, 4 and 5 (NP_062816.2; NP_001153602.1; NP_001153604.1; NP_1153605.1; NP_001153606.1).
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
KCNQ5
Peptide with sequence C-ESTDALSLPHVKLK corresponding to C-Terminus according to NP_001153602.1, NP_001153604.1, NP_1153605.1, NP_001153606.1.
Immunogen affinity purified
RUO
Primary
56479
Canine, Human, Mouse, Porcine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only