Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KY Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523914
Description
KY Polyclonal antibody specifically detects KY in Human, Mouse samples. It is validated for Western Blot.Specifications
KY | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8C8H8, Q8NBH2 | |
KY | |
KLH conjugated synthetic peptide between 104-133 amino acids from the N-terminal region of human KY | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
KY | |
CD1; D9Mgc44e; ky; Kyphoscoliosis peptidase | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
16716, 339855 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction