Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Ficolin Monoclonal Antibody (16), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific ABS0051602
Description
ABS 005-16-02 detects L-Ficolin from human samples. ABS 005-16-02 has been successfully used in ELISA and Western blot procedures. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
L-Ficolin Monoclonal antibody specifically detects L-Ficolin in Human samples. It is validated for ELISA, Western Blot.Specifications
L-Ficolin | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Q15485 | |
FCN2 | |
Purified recombinant protein corresponding to human L-Ficolin and full-length L-Ficolin (Ficolin-2). | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
16 | |
Unconjugated | |
FCN2 | |
37 kDa elastin-binding protein; Collagen/fibrinogen domain-containing protein 2; EBP-37; FCN2; Fcnb; FCNL; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin 2; ficolin B; ficolin-2; Ficolin-B; ficolin-beta; Hucolin; L-ficolin; P35; RP11-263F14.2; Serum lectin p35 | |
Mouse | |
Protein A/G | |
RUO | |
2220 | |
4° C, store in dark | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction