Learn More
Listeria monocytogenes, HRP, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA185652
Catalog No. PIPA185652
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
The genus Listeria comprises six species: L. monocytogenes, L. innocua, L. welshimeri, L. seeligeri, L. ivanovii and L. grayi. Listeria monocytogenes, the most commonly isolated pathogenic member, is associated with a wide spectrum of human and animal diseases. In the smear from the original tissue, L. monocytogenes may appear as gram-positive coccobacilli that may be confused with Streptococcus agalactiae (group B), enterococci, or Corynebacterium spp. Listeria is differentiated from streptococci by a positive catalase test.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Listeria monocytogenes
Polyclonal
HRP
Rabbit
Purified
RUO
Bacteria
Antibody
IgG
ELISA, Immunohistochemistry (Frozen), Western Blot
1mg/mL
L. monocytogenes
Listeria monocytogenes; ATCC strain 43251
1 mL
Primary
Store at 4°C or Store at -20°C if preferred
Purified
Product Suggestions
Product Suggestions