Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Myc Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130045

Catalog No. PA130045


Add to cart

Description

Description

PA1-30045 detects l-Myc from human samples. PA1-30045 has been successfully used in immunohistochemistry (paraffin tissue), Western blot applications. The PA1-30045 immunogen is: Synthetic peptide derived from residues 105 - 154 of human l-Myc.

Cellular DNA-binding proteins encoded by the c-myc genes. They are normally involved in nucleic acid metabolism and in mediating the cellular response to growth factors. Elevated and deregulated (constitutive) expression of c-myc proteins can cause tumorigenesis. L-Myc gene ontology annotations include DNA binding function, DNA-binding transcription factor activity, RNA polymerase II-specific action, protein dimerization activity, and regulation of inner ear auditory receptor cell differentiation.
Specifications

Specifications

l-Myc
Polyclonal
Unconjugated
MYCL
Liquid
MYCL
Synthetic peptide derived from residues 105 - 154 of human l-Myc.
Protein A
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P12524
LMYC, MYCL, bHLHe38
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4610
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit