Learn More
CD62L Sheep anti-Human, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Supplier:  Thermo Scientific PA175241
Catalog No. PIPA175241
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
PA1-75241 specifically detects L-Selectin. PA1-75241 has been successfully used in ELISA applications. The PA1-75241 immunogen is Recombit L-Selectin.
This gene encodes a cell surface adhesion molecule that belongs to a family of adhesion/homing receptors. The encoded protein contains a C-type lectin-like domain, a calcium-binding epidermal growth factor-like domain, and two short complement-like repeats. The gene product is required for binding and subsequent rolling of leucocytes on endothelial cells, facilitating their migration into secondary lymphoid organs and inflammation sites. Single-nucleotide polymorphisms in this gene have been associated with various diseases including immunoglobulin A nephropathy. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD62L
Polyclonal
SELL
SELL, LAM-1, LAM1, LECAM1, LNHR, LSEL, LYAM1, Leu-8, Lyam-1, PLNHR, TQ1, hLHRc
Sheep
Affinity chromatography
RUO
6402
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ELISA
Unconjugated
P14151
SELL
Recombit human L-Selectin
500 μg
Primary
Human
Antibody
IgG
Recommendations
Recommendations