Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Thyroxine T4 Mouse anti-Chemical, Clone: T4YCH, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MIT0501
Description
MIT0501 targets in ELISA, Western Blot, IP, and RIA applications and shows reactivity with Chemical samples. MIT0501 was formerly sold as a Seradyn product.
Levothyroxine, also L-thyroxine, synthetic T4, or 3,5,3′,5′-tetraiodo-L-thyronine, is a synthetic form of thyroxine (thyroid hormone), used as a hormone replacement for patients with thyroid problems. The natural hormone is chemically in the chiral L-form, as is the pharmaceutical agent. Dextrothyroxine (D-thyroxine) briefly saw research as an anticholesterol agent but was pulled due to cardiac side-effects.Specifications
L-Thyroxine T4 | |
Monoclonal | |
1 mg/mL | |
L-thyroxine; Levothyroxine; Thyroxine; Synthroid; C15H11I4NO4 | |
Thyroxine-BSA | |
1 mg | |
Primary | |
Antibody | |
IgG2b |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
T4YCH | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid | |
Chemical |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction