Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-L3MBTL2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132621

Catalog No. NBP132621


Add to cart

Description

Description

L3MBTL2 Polyclonal antibody specifically detects L3MBTL2 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation.
Specifications

Specifications

L3MBTL2
1.0mg/mL
Western Blot 1:500-1:3000, Chromatin Immunoprecipitation, Immunoprecipitation 1:500-1:1000
Affinity Purified
L3MBTL2
Recombinant protein encompassing a sequence within the center region of human L3MBTL2. The exact sequence is proprietary.
79 kDa
100 ul
Apoptosis, Cancer, Tumor Suppressors
Primary
83746
Human, Mouse
ChIP assay, Immunoprecipitation, Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
dJ756G23.3, DKFZP761I141, H-l(3)mbt-l, H-l(3)mbt-like protein 2, l(3)mbt-like 2 (Drosophila), L(3)mbt-like protein 2, L3MBT, lethal(3)malignant brain tumor-like protein 2
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only