Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L3MBTL4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA529477

Catalog No. PIPA529477


Add to cart

Description

Description

PA5-29477 targets L3MBTL4 in WB applications and shows reactivity with Human and mouse samples. The PA5-29477 immunogen is recombit fragment corresponding to a region within amino acids 199 and 494 of Human L3MBTL4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility.
Specifications

Specifications

L3MBTL4
Polyclonal
Unconjugated
L3MBTL4
Liquid
L3MBTL4
Recombit fragment corresponding to a region within amino acids 199 and 494 of Human L3MBTL4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q8NA19, B1B1A0
HsT1031, H-l(3)mbt-like protein 4, l(3)mbt-like protein 4, lethal(3)maligt brain tumor-like protein 4
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
320858, 91133
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit