Learn More
La Crosse Virus Mouse anti-Virus, Clone: 4A5.5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110802
Catalog No. 01-671-831
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10802 detects La Crosse Virus. MA1-10802 has been successfully used in ELISA, Western blot, immunofluorescence, immunoprecipitation, and neutralization applications. The MA1-10802 immunogen is La Crosse Virus grown on E6 Vero cells and purified by sucrose gradient centrifugation.
Specifications
Specifications
La Crosse Virus
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
LCV
La Crosse Virus grown on E6 Vero cells and purified by sucrose gradient centrifugation.
Protein G
RUO
Antibody
Monoclonal
ELISA, Immunofluorescence, Immunoprecipitation, Neutralization, Western Blot
4A5.5
Unconjugated
Liquid
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Virus