Learn More
La Crosse Virus Mouse anti-Virus, Clone: 9E7.2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110803
Catalog No. 01-671-832
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10803 detects La Crosse Virus. MA1-10803 has been successfully used in ELISA, Western blot, immunofluorescence, immunoprecipitation, and viral neutralization applications. The MA1-10803 immunogen is La Crosse Virus grown on E6 Vero cells and purified by sucrose gradient centrifugation.
Specifications
Specifications
La Crosse Virus
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
LCV
La Crosse Virus grown on E6 Vero cells and purified by sucrose gradient centrifugation.
Purified
RUO
Antibody
Monoclonal
ELISA, Immunofluorescence, Immunoprecipitation, Neutralization, Western Blot
9E7.2
Unconjugated
Liquid
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Virus