Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
La Protein Mouse, Unlabeled, Clone: 44, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610905
Description
La was first found to bind autoantibodies from individuals with Sjogren's syndrome and systemic lupus erythematosus (SLE). La is a transcription factor for RNA polymerase III (Pol III). It participates in the termination of Pol III-media
Specifications
La Protein | |
Monoclonal | |
250μg/mL | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
IgG1 |
Immunofluorescence, Western Blot | |
44 | |
Unconjugated | |
Human La Protein aa. 179-289 | |
150 μg | |
Cell Biology | |
Canine, Human, Rabbit, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction