Learn More
LacI Mouse anti-All, Clone: 9A5, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05503
Catalog No. 05-503-MI
Add to cart
Description
Description
Detect LacI using this Anti-LacI Antibody, clone 9A5 validated for use in western blot, immunocytochemistry, immunohistochemistry.
Specifications
Specifications
LacI
9A5
Lot specific (please refer to your antibody associated certificate of analysis).
Purified
TrpE fusion to full-length E. coli LacI
Protein G chromatography
−20°C for 24 months.
Monoclonal
Immunocytochemistry, Immunohistochemistry, Western Blot
Unconjugated
NP_414879.3
Murine
IgG1
200μg
Primary
All