Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactate Dehydrogenase A/LDHA Rabbit anti-Human, Mouse, Porcine, Bovine, Biotin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP148336B

Catalog No. NBP148336B


Add to cart

Description

Description

Lactate Dehydrogenase A/LDHA Polyclonal antibody specifically detects Lactate Dehydrogenase A/LDHA in Human, Mouse, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

Lactate Dehydrogenase A/LDHA
Biotin
PBS with 0.05% Sodium Azide
Affinity Purified
LDHA
Synthetic peptide made to a C-terminal portion of the human Lactate Dehydrogenase A protein (within residues 280-332). [Swiss-Prot# P00338]
36 kDa
0.1 ml
Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Neuroscience
Primary
3939
Bovine, Human, Mouse, Porcine
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
P00338
Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human and mouse. Predicted to react with bovine and porcine based on 100% sequence homology. Bovine data from customer review.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only