Learn More
LDHA Goat anti-Rabbit, Biotin, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA126911
Catalog No. PA126911
Add to cart
Description
Description
PA1-26911 detects Lactate Dehydrogenase from rabbit samples. PA1-26911 is expected to cross react with a wide range of species due to sequence homology. PA1-26911 has been successfully used in ELISA, immunoprecipitation and Western blot procedures. The PA1-26911 immunogen is purified, full length, native Lactate Dehydrogenase. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
Specifications
LDHA
Polyclonal
Biotin
LDHA
Liquid
Goat
IgG
1 mg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100009107
ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
10 mg/mL
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% sodium azide; pH 7.2
P13491
LDHA
Purified, full length, native Lactate Dehydrogenase.
IgG fraction
RUO
Antibody
Polyclonal
Rabbit