Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Lactoperoxidase, DyLight 650, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB11061572C

Catalog No. NB11061572C


Add to cart

Description

Description

Lactoperoxidase Polyclonal specifically detects Lactoperoxidase in Human, Bovine, Primate samples. It is validated for Western Blot.
Specifications

Specifications

Lactoperoxidase
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
LPO
Synthetic peptide made to an internal portion of the human Lactoperoxidase protein (within residues 300-400). [Swiss-Prot# P22079]
100 kDa
0.1 ml
Cancer, Immunology, Virology Bacteria and Parasites
Primary
4025
Human, Bovine, Primate
Western Blot
Western Blot
P22079
EC 1.11.1, lactoperoxidase, MGC129990, MGC129991, Salivary peroxidase, SAPX, SPOEC 1.11.1.7
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, monkey and bovine. Immunogen has 81% homology to sheep and 73% homology to mouse and rat.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only