Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoperoxidase Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11061572
Description
Lactoperoxidase Polyclonal specifically detects Lactoperoxidase in Human, Bovine, Primate samples. It is validated for Western Blot.Specifications
Lactoperoxidase | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.1% Sodium Azide | |
EC 1.11.1, lactoperoxidase, MGC129990, MGC129991, Salivary peroxidase, SAPX, SPOEC 1.11.1.7 | |
Rabbit | |
100 kDa | |
0.1 mL | |
Cancer, Immunology, Virology Bacteria and Parasites | |
4025 | |
Human, Bovine, Primate | |
IgG |
Western Blot | |
1.7 mg/mL | |
Western Blot 0.5-2 ug/ml | |
P22079 | |
LPO | |
Synthetic peptide made to an internal portion of the human Lactoperoxidase protein (within residues 300-400). [Swiss-Prot# P22079] | |
Affinity Purified | |
RUO | |
Primary | |
Human, monkey and bovine. Immunogen has 81% homology to sheep and 73% homology to mouse and rat. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction