anti-LAIR1, Polyclonal, Abnova

Mouse Polyclonal

Manufacturer: Abnova Corporation H00003903B01

 View more versions of this product

Catalog No. 89-005-686

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Regulatory Status RUO
    Whole Molecule Y
    Conjugate Unlabeled
    Cross Reactivity Human
    Format Serum
    Host Species Mouse
    Immunogen LAIR1 (AAH27899, 1 a.a. ∽288 a.a) full-length human protein.
    Quantity 50μL
    Primary or Secondary Primary
    Monoclonal or Polyclonal Polyclonal
    Antigen leukocyte-associated immunoglobulin-like receptor 1

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit