Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin A/C Mouse, Unlabeled, Clone: 14, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612162

 View more versions of this product

Catalog No. BDB612162


Add to cart

Description

Description

The nuclear envelope (NE) is a specialized extension of the ER that contains numerous pore complexes interconnected with the nuclear lamina. The nuclear lamina composes the structural framework for the NE and serves as a chromatin anchor site, thus playing a major role in interphase nuclear organization. The major component of nuclear lamina are intermediate filament-like proteins called lamins. In mammalian somatic cells, there are three major lamins, A, B1, and C, and two minor lamins, B2 and A10. A-type lamins (A, A10, and C) are encoded by a
single gene and are produced by alternative splicing, while B-type lamins (B1 and B2) are encoded by separate genes. B-type lamins are found in all nucleated somatic cells, while the expression of A-type lamins are developmentally regulated. Mice lacking lamin A show no overt abnormalities until postnatal development when perturbations in nuclear envelop structure correlate with the appearance of muscular dystrophy. In addition, lamin A is mutated in lipodystrophy, a disorder characterized by reduction in subcutaneous adipose tissue. Thus, lamin A and C may be important for nuclear envelope formation during postnatal cell differentiation.

Immunofluorescence, Western Blotting

Specifications

Specifications

Lamin A/C
14
Unconjugated
Affinity Purified
Human Lamin A/C aa. 398-490
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.