Learn More
Lamin A/C Mouse anti-Human, Mouse, Rat, Canine, Chicken, Clone: 14, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05714
Catalog No. 05-714-MI
Add to cart
Description
Description
Anti-Lamin A/C Antibody, clone 14 detects level of Lamin A/C and has been published and validated for use in IC and western blotting.
Specifications
Specifications
Lamin A/C
14
Purified mouse monoclonal IgG1 in buffer containing 50% storage buffer (20mM sodium phosphate, pH 7.5, 0.15MNaCl, 1mg/mL BSA, 0.09% sodium azide) and 50% glycerol. Store at −20°C.
Purified
Murine
IgG
50μg
Primary
Canine, Chicken, Human, Murine, Rat
Immunocytochemistry, Western Blot
Unconjugated
NM_005572
LMNA, HGPS, EMD2, FPLD, CDCD1, LDP1, LGMD1B, IDC, CMT2B1, LMNC, FPL, PRO1, Lamin-A/C, LMN1, CMD1A, LFP, CDDC
Peptide from human lamin A/C corresponding to amino acids 398 to 490.
Protein G purfied
−20°C for up to 12 months.
Monoclonal