Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin A + C Rabbit anti-Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP225153SS

 View more versions of this product

Catalog No. NBP225153SS

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Lamin A + C Polyclonal antibody specifically detects Lamin A + C in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Lamin A + C
Unconjugated
Whole antisera with 10mM Sodium Azide
Antisera
LMNA
Recombinant full length purified Lamin A + C from E. coli
Unpurified
RUO
Primary
4000
Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Western Blot 1:5000, Immunohistochemistry 1:5000, Immunocytochemistry/Immunofluorescence 1:5000
P02545
CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32
Rabbit
IgG
0.025 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Bovine, Porcine, Mouse, Rat and other Mammals. Other species have not been tested.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only