Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin B1/B2 Mouse anti-Bovine, Human, Rat, Clone: X223, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA15822

Catalog No. 01-674-929


Add to cart

Description

Description

MA1-5822 detects Lamin B1 & B2 in human, bovine, and rat samples, and Lamin B2 in mouse. MA1-5822 has been successfully used in immunofluorescence and Western blot procedures. The MA1-5822 immunogen is nuclear pore complex-lamina fraction of XLKE-A6 cells. Store at 4°C of one month, or in small aliquots at -20°C for longer periods, avoiding freeze/thaw cycles.

This gene is a member of the transferrin family of genes and its protein product is found in the secondary granules of neutrophils. The protein is a major iron-binding protein in milk and body secretions with an antimicrobial activity, making it an important component of the non-specific immune system. The protein demonstrates a broad spectrum of properties, including regulation of iron homeostasis, host defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation and protection against cancer development and metastasis.
Specifications

Specifications

Lamin B1/B2
Monoclonal
Unconjugated
LMNB1
Liquid
LMNB1, LMNB2
Nuclear pore complex-lamina fraction of Xenopus laevis (XLKE-A6 cells)
2 mL
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
116685, 299625, 4001, 516326, 540643, 84823
Immunofluorescence, Western Blot
X223
tissue culture supernatant with 0.09% sodium azide
P20700, Q03252, P70615
B type lamin
Mouse
IgG1
RUO
Antibody
Monoclonal
Bovine, Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit