Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Lamin B1, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP142594

Catalog No. NBP142594

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Lamin B1
LYOPH
Western Blot 0.1-0.5ug/ml, Immunohistochemistry 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 1:10 - 1:500
P20700
ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, MGC111419
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Reconstitute with 0.2 ml distilled water to a final concentration of 0.5 mg/ml.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3. with 0.05mg Thimerosal
Affinity Purified
LMNB1
A synthetic peptide corresponding to a sequence at the C-terminal of human Lamin B,different from the related rat and mouse sequence by three amino acids.
66.4 kDa
0.1 mg
Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
Primary
4001
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only