Learn More
Lamin A/C Mouse anti-Human, Mouse, Clone: Jol5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA15821
Catalog No. 01-674-928
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-5821 detects an un-phosphorylated form of lamin A/C and the epitope is mapped to the N-terminal head domain of both proteins (aa 1-171). It reacts with mouse myoblasts (C2C12) in Western blot applications. Other cell/tissue types have not been tested in Western blot. Detects a band of approximately 70 kDa (Lamin A) and 60kDa (Lamin C).
Nuclear lamins form a network of intermediate-type filaments at the nucleoplasmic site of the nuclear membrane. Two main subtypes of nuclear lamins can be distinguished, i.e. A-type lamins and B-type lamins. The A-type lamins comprise a set of three proteins arising from the same gene by alternative splicing, i.e. lamin A, lamin C and lamin Adel 10, while the B-type lamins include two proteins arising from two distinct genes, i.e. lamin B1 and lamin B2. Recent evidence has revealed that mutations in A-type lamins give rise to a range of rare but domit genetic disorders, including Emery-Dreifuss muscular dystrophy, dilated cardiomyopathy with conduction-system disease and Dunnigan-type familial partial lipodystrophy. In addition, the expression of A-type lamins coincides with cell differentiation and as A-type lamins specifically interact with chromatin, a role in the regulation of differential gene expression has been suggested for A-type lamins.
Specifications
Specifications
Lamin A/C
Monoclonal
Unconjugated
LMNA
Liquid
LMNA
Recombit bacterially expressed full length protein
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
16905, 4000
Immunocytochemistry, Immunofluorescence, Western Blot
Jol5
tissue culture supernatant with 0.1% sodium azide
P02545, P48678
Prelamin-A/C, Lamin-A/C, 70 kDa lamin, Renal carcinoma antigen NY-REN-32, LMNA, LMN1
Mouse
IgG2
RUO
Antibody
Monoclonal
Human, Mouse