Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin C Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA15827

Catalog No. 01-674-977


Add to cart

Description

Description

PA1-5827 detects Lamin C in human, rat and mouse samples. PA1-5827 has been successfully used in immunofluorescence, Western blot and immunohistochemistry procedures. PA1-5827 reacts strongly in immunoblotting with fragments corresponding to peptide 319-572aa of Lamin C. When tested against nuclear protein extracts from HeLa cells it reacts with a single band migrating at 65 kDa. The PA1-5827 immunogen is a synthetic peptide corresponding to the N-terminal residues K-HHVSGSRR of human lamin C, coupled to KLH.

Nuclear lamins form a network of intermediate-type filaments at the nucleoplasmic site of the nuclear membrane. Two main subtypes of nuclear lamins can be distinguished, i.e. A-type lamins and B-type lamins. The A-type lamins comprise a set of three proteins arising from the same gene by alternative splicing, i.e. lamin A, lamin C and lamin Adel 10, while the B-type lamins include two proteins arising from two distinct genes, i.e. lamin B1 and lamin B2. Recent evidence has revealed that mutations in A-type lamins give rise to a range of rare but domit genetic disorders, including Emery-Dreifuss muscular dystrophy, dilated cardiomyopathy with conduction-system disease and Dunnigan-type familial partial lipodystrophy. In addition, the expression of A-type lamins coincides with cell differentiation and as A-type lamins specifically interact with chromatin, a role in the regulation of differential gene expression has been suggested for A-type lamins.
Specifications

Specifications

Lamin C
Polyclonal
PBS with 0.1% sodium azide
P02545, P48678, P48679
C type lamin
Rabbit
IgG
200 μL
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
16905, 4000, 60374
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LMNA
Liquid
LMNA
A peptide made from the last 8 amino acids of lamin C, including an N-terminal lysine as a linker (KHHVSGSRR), coupled to KLH
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit