Learn More
MilliporeSigma™ anti-Laminin γ2 (Laminin 5), Clone: D4B5, ™ (Chemicon™)
Manufacturer:  MilliporeSigma™ MAB19562/DEL
Catalog No. 50-174-998
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Laminins are basement membrane, extracellular matrix glycoproteins that are composed of alpha, beta and gamma heterotrimeric chains. Different combinations of five alpha chains, three beta chains, and two gamma chains form at least 12 laminin isoforms. Laminin-5 (LN5), which consists of laminin alpha3, beta3 and gamma2 chains, is a highly conserved component of anchoring filaments underlying the hemidesmosome structure of epidermal keratinocytes. Mutations or deletion in the LN5 genes (LAMA3, LAMB3, and LAMC2) is associated with epidermolysis bullosa, a lethal skin blistering disease. LN5 may stabilize the epidermal/dermal junction of skin through binding with integrins alpha3/beta1, alpha6/beta4 and type VII collagen and has a critical role in the anchorage and locomotion of a wide variety of cell types, including epithelial cells, fibroblasts, neurons and leukocytes. Compared to fibronectin, collagen, or vitronectin, cells of epithelial origin will adhere to Laminin-5 faster and will spread to a larger extent. Recently, Laminin-5 is attracting attention as a substrate for carcinoma cells that may stimulate migration, since it has been detected at the leading edge of invasive cancer tissue in several types of human carcinomas (Niki, et al., 2002, Lohi, et al., 2001).
ELISA, Immunohistochemistry (Paraffin), Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
Laminin γ2 (Laminin 5)
Unconjugated
Amino acid residues 382-608 of human laminin γ 2 chain (III domain) expressed as a GST fusion protein
RUO
Primary
Semi-purified
Monoclonal
Murine
100 μg
Cell Structure
Human
IgG1