Learn More
anti-Laminin alpha 4, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP132159
Catalog No. NBP132159
Add to cart
Description
Description
Laminin alpha 4 Polyclonal antibody specifically detects Laminin alpha 4 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Laminin alpha 4
0.74mg/mL
Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
LAMA4
Recombinant protein encompassing a sequence within the C-terminus region of human Laminin alpha 4. The exact sequence is proprietary.
203 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in Mouse. Not yet tested in any other species.
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
DKFZp686D23145, LAMA3LAMA4*-1, laminin alpha 4 chain, laminin subunit alpha-4, laminin, alpha 4, Laminin-14 subunit alpha, Laminin-8 subunit alpha, Laminin-9 subunit alpha
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
3910
Mouse