Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin B2 Mouse, Unlabeled, Clone: 22, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610722

Catalog No. BDB610722


Add to cart

Description

Description

The basal lamina contains Collagen Type IV, proteoglycans, and glycoproteins. Laminin is a high molecular weight ( ∽850 kDa) oligomer, consisting of three different chains (A, B1, and B2) joined by disulfide bonds. The overall structure predicts two helical domains (I&II) at the COOH-terminal, cysteine-rich repeats (III&V), and two globular domains (IV&VI). Domains IV and VI are the binding sites for collagen and heparan sulfate, respectively. Several isoforms have been identified for the genes of each chain. Laminin B2 is 1576 amino acids plus a 33 amino acid signal peptide, 14 glycosylation sites, and 12 cysteine repeat domains. The expression of the Laminin subunits is detected rapidly during development and tissue regeneration. Attachment of cells to the basal lamina affects mitogenesis, motility, and differentiation.
Specifications

Specifications

Laminin B2
22
Unconjugated
Affinity Purified
Human Laminin B2 aa. 506-691
50μg
Cell Biology
Primary
Human, Rabbit, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit