Learn More
Laminin Rabbit anti-Human, Mouse, Ovine, Rabbit, Rat, Biotin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522900
Catalog No. PIPA522900
Add to cart
Description
Description
This antibody is pan-specific and reacts well with all Laminin isoforms tested: Laminin-1 (alpha-1, beta-1, and gamma-1) and Laminin-2 (alpha-2, beta-1, and gamma-1). The antibody functionally inhibits Laminin in mouse and rat. It binds to Laminin and inhibits most, if not all, of its cell adhesion and growth promotive properties. Immunostaining is enhanced by antigen retrieval with pepsin, especially paraffin tissue. Suggested positive control: basement membranes (blood vessels of any tissue, tumors, nerve).
Laminin antibodies are widely used to label blood vessels and basement membranes.
Specifications
Specifications
Laminin
Polyclonal
Biotin
LAMA1
Liquid
LAMA1, LAMA2, LAMB1, LAMC1
Laminin isolated from mouse EHS tumor.
IgG fraction
RUO
Antibody
Polyclonal
Human, Mouse, Ovine, Rabbit, Rat
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
1.0 mg/mL
PBS with 0.05% sodium azide
P24043, P11047, P07942, P19137, P25391
LAMA, S-LAM-alpha, S-LAM alpha, S-laminin subunit alpha, laminin A chain, laminin subunit alpha-1, laminin-3 subunit alpha
Rabbit
IgG
100 μL
4° C, store in dark
Primary
284217, 316758, 3915, 16772, 3912, 100357482, 3908, 101102692