Learn More
Laminin gamma-1 Mouse anti-Bovine, Guinea Pig, Human, Porcine, Rabbit, Rat, Clone: C4, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA126516
Catalog No. MA126516
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-26516 detects Laminin S from human, rat, bovine, porcine, guinea pig, and rabbit samples. MA1-26516 has been successfully used in Western blot and immunoprecipitaton procedures. Positive control:LS174T cells. The MA1-26516 immunogen is bovine anterior lens capsule.
Specifications
Specifications
Laminin gamma-1
Monoclonal
0.2 mg/mL
10mM PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4
P11047
LAMC1
Bovine anterior lens capsule.
Protein A
RUO
Antibody
Monoclonal
Bovine, Guinea Pig, Human, Porcine, Rabbit, Rat
Immunoprecipitation, Western Blot
C4
Unconjugated
LAMC1
Liquid
Mouse
IgG1
500 μL
4° C, do not freeze
Primary
100734278, 532572, 3915, 100514785, 117036, 103346447