Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Langerin Mouse anti-Human, FITC, Clone: 343828, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523638

Catalog No. PIMA523638

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

This antibody does not cross-react with recombit human (rh) Granzyme A, rhGranzyme H, recombit mouse (rm) Granzyme B, rmGranzyme C, rmGranzyme D, or rmGranzyme G.

The protein encoded by this gene is expressed only in Langerhans cells which are immature dendritic cells of the epidermis and mucosa. It is localized in the Birbeck granules, organelles present in the cytoplasm of Langerhans cells and consisting of superimposed and zippered membranes. It is a C-type lectin with mannose binding specificity, and it has been proposed that mannose binding by this protein leads to internalization of antigen into Birbeck granules and providing access to a nonclassical antigen-processing pathway.
Specifications

Specifications

Langerin
Monoclonal
FITC
CD207
Liquid
CD207
Mouse myeloma cell line NS0-derived recombit human Langerin/CD207 Tyr64-Pro328
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
343828
Buffered saline solution with BSA and 0.09% sodium azide
Q9UJ71
CLEC4K, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, CD207 antigen, langerin, Langerhans cell specific c-type lectin
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
50489
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit