Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAR Mouse, Unlabeled, Clone: 7, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610350

 View more versions of this product

Catalog No. BDB610350


Add to cart

Description

Description

The Leukocyte common Antigen-Related (LAR) receptor protein tyrosine phosphatase (RPTP) is one of approximately 20 known, distinct transmembrane RPTPs. LAR consists of an extracellular region with three Ig-like and eight fibronectin type III-like (FNIII) domains and a cytoplasmic region containing two tyrosine phosphatase domains typical of RPTPs. The Ig and FNIII domains of LAR show some sequence similarity to domains in the neural cell adhesion molecule N-CAM, a neural surface glycoprotein which mediates cell adhesion and neurite outgrowth. Studies suggest that LAR regulates neurite outgrowth and pathfinding during neural development. N-CAM and other cell adhesion molecules that affect neurite outgrowth, such as L1, TAG-1, and fasciculin II, lack intrinsic kinase or phosphatase activity. Thus, LAR is part of a distinct class of proteins with both cell adhesion and tyrosine phosphatase functions. LAR transcripts are alternatively spliced and that splicing is regulated during development. It is postulated that the various spliced forms of LAR each have specialized functions in the nervous system and play a role in neural development and regeneration.

Specifications

Specifications

LAR
7
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Leukocyte common Antigen-Related
Affinity Purified
Aqueous buffered solution containing BSA
50μg
Cell Biology
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.