Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LARGE Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP149865
Description
LARGE Polyclonal specifically detects LARGE in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
LARGE | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Acetylglucosaminyltransferase-like 1A, EC 2.4, glycosyltransferase-like protein LARGE1, KIAA0609acetylglucosaminyltransferase-like protein, LARGE1, like-acetylglucosaminyltransferase, like-glycosyltransferase, MDC1D, MDDGA6, MDDGB6 | |
Goat | |
Affinity Purified | |
RUO | |
9215 | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:2000 | |
NP_004728.1 | |
LARGE | |
Peptide with sequence C-SEADVNSENLQKQ corresponding to internal region according to NP_004728.1. | |
0.1 mg | |
Primary | |
Reported variants represent identical protein: NP_004728.1, NP_598397.1 | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction