Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LARP6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524317

Catalog No. PIPA524317


Add to cart

Description

Description

LARP6 is a member of the La ribonucleoprotein domain family, which all contain one characteristic La motif. LARP6 shuttles between the cytoplasm and nucleus. It is proposed that members of this family play a role in translation and mRNA stability.
Specifications

Specifications

LARP6
Polyclonal
PBS with 0.09% sodium azide
Q9BRS8, Q8BN59
ACHN, acheron, death-associated LA motif protein, la-related protein 6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55323, 67557
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LARP6
Liquid
LARP6
KLH conjugated synthetic peptide between 7-35 amino acids from the N-terminal region of human LARP6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit