Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-LARS, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232209

Catalog No. NBP232209


Add to cart

Description

Description

LARS Polyclonal antibody specifically detects LARS in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

LARS
1.0 mg/mL
Western Blot 1:1000 - 1:5000, Immunohistochemistry
NP_064502.9
cytoplasmic leucyl-tRNA synthetase, FLJ21788, hr025Cl, HSPC192, KIAA1352, LARS1, leucine translase, leucine tRNA ligase 1, cytoplasmic, leucine-tRNA ligase, leucine--tRNA ligase, leucyl-tRNA synthetase, leucyl-tRNA synthetase, cytoplasmic, LEURS, LEUS, LRS, PIG44
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Western Blot, Immunohistochemistry
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
LARS
The immunogen this antibody was made to, maps to a region between residue 250 to 300 of human Leucyl-tRNA Synthetase using the numbering given in entry NP_064502.9 (GeneID 51520).
Immunogen affinity purified
RUO
Primary
51520
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only