Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LARS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514309

Catalog No. PIPA514309

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

LARS, a cytosolic leucine-tRNA synthetase, a member of the class I aminoacyl-tRNA synthetase family. This enzyme catalyzes the ATP-dependent ligation of L-leucine to tRNA(Leu). It is found in the cytoplasm as part of a multisynthetase complex and interacts with the arginine tRNA synthetase through its C-terminal domain.
Specifications

Specifications

LARS
Polyclonal
PBS with 0.09% sodium azide
Q9P2J5
Leucyl-tRNA synthetase, cytoplasmic; Leucine--tRNA ligase; LeuRS; LARS
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51520
Immunohistochemistry, Western Blot
Unconjugated
LARS
Liquid
LARS
KLH conjugated synthetic peptide between 1118-1145 amino acids from the C-terminal region of human LARS
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit