Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lass4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112927

Catalog No. PIPA112927


Add to cart

Description

Description

A suggested positive control for this product is lung or embryo lysate.

It is thought that Lass4 may be either a bona fide (dihydro)ceramide synthase or a modulator of its activity. When overexpressed in cells is involved in the production of sphingolipids containing different fatty acid donnors (N-linked stearoyl- (C18) or arachidoyl- (C20) ceramides) in a fumonisin B1-independent manner.
Specifications

Specifications

Lass4
Polyclonal
Unconjugated
CERS4
Liquid
CERS4
Synthetic peptide corresponding to human Lass4 protein.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.08% sodium azide
Q9HA82
CerS4, FLJ12089, Trh1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
79603
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit