Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAT Mouse anti-Human, Mouse, Clone: 11B.12, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05770
Description
This Anti-LAT Antibody, clone 11B.12 is validated for use in immunoprecipitation, western blotting for the detection of LAT.Specifications
LAT | |
Monoclonal | |
Unconjugated | |
NM_014387 | |
Mouse | |
Protein G purfied | |
Primary | |
−20°C for 24 months. | |
IgG1 |
Immunoprecipitation, Western Blot | |
11B.12 | |
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% | |
LAT, LAT1, pp36, p36-38 | |
GST fusion protein corresponding to residues 31-233 of human LAT, cytoplasmic domain | |
100 μL | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction