Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Latent TGF-beta bp1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 9 publications
$184.44 - $1184.00
Specifications
Antigen | Latent TGF-beta bp1 |
---|---|
Clone | 35409 |
Dilution | Western Blot 1 ug/mL, Immunoaffinity Purification |
Applications | Western Blot, Affinity Purification |
Classification | Monoclonal |
Description
Latent TGF-beta bp1 Monoclonal specifically detects Latent TGF-beta bp1 in Human samples. It is validated for Western Blot, Immunoaffinity Purification.Specifications
Latent TGF-beta bp1 | |
Western Blot 1 ug/mL, Immunoaffinity Purification | |
Monoclonal | |
Mouse | |
Human | |
Latent TGFbeta bp1, latent transforming growth factor beta binding protein 1, latent-transforming growth factor beta-binding protein 1, LTBP1, LTBP-1, MGC163161, TGF-beta1-BP-1, Transforming growth factor beta-1-binding protein 1 | |
LTBP1 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
35409 | |
Western Blot, Affinity Purification | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
4052 | |
Human platelet-derived large latent TGF-beta 1 complex | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human LTBP-1 from the large latent TGF-beta 1 complex. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title