Learn More
Latexin Rabbit anti-Human, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523239
Catalog No. PIPA523239
Add to cart
Description
Description
Latexin, or tissue carboxypeptidase inhibitor (TCI), is the only known mammalian carboxypeptidase inhibitor. It is often related to a human putative tumor suppressor protein, TIG1. Latexin was first identified as a marker of neurons in the lateral neocortex of the developing mammalian brain. Latexin plays a role in the modulation of sensory perception and also functions in inflammation and innate immune pathways. The protein also contributes to the pathological disorders acute pancreatitis and lung inflammation. Latexin is expressed in heart, prostate, ovary, kidney, pancreas, colon and brain. The human latexin gene is assigned to chromosomal region 3q25.32.
Specifications
Specifications
Latexin
Polyclonal
whole serum with 50% glycerol and 0.025% sodium azide
Q9BS40, Q64361
ECI, TCI, MUM, endogenous carboxypeptidase inhibitor, tissue carboxypeptidase inhibitor
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Rat
Immunohistochemistry (Paraffin)
Unconjugated
LXN
Liquid
LXN
Synthetic peptide (70 CTAEVLYPSTGQETAPE 86) of Latexin protein.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56925, 59073