Learn More
LBX1 Mouse anti-Human, Clone: 2B2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA521116
Catalog No. PIMA521116
Add to cart
Description
Description
Peptide Sequence: TNHQIYELEK RFLYQKYLSP ADRDQIAQQL GLTNAQVITW FQNRRAKLKR DLEEMKADVE SAKKLGPSGQ MDIVALAELE QNSEATA
This gene and the orthologous mouse gene were found by their homology to the Drosophila lady bird early and late homeobox genes. In the mouse, this gene is a key regulator of muscle precursor cell migration and is required for the acquisition of dorsal identities of forelimb muscles.
Specifications
Specifications
LBX1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P52954
Ladybird homeobox 1, HPX-6, HPX6, LBX1H, homeobox
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10660
ELISA, Western Blot
2B2
Unconjugated
LBX1
Liquid
LBX1
LBX1 (NP_006553.2, 133 a.a. ∽ 219 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human