Learn More
LC3B, DyLight® 550, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522713
Catalog No. PIPA522713
Add to cart
Description
Description
This antibody has 92% homology with mouse, 91% with bovine, and 84% with xenopus. Reacts weaker with mouse than with human samples
LC3B (Autophagy Marker Light Chain 3B, MAP1A/MAP1B LC3 B) in humans, is encoded by the gene MAP1LC3B (Microtubule-associated proteins 1A/1B light chain 3B). LC3B is associated with microtubule assembly and important is in neurogenesis. Recent studies indicate that LC3B plays a vital role in autophagy, a process that involves the bulk degradation of cytoplasmic component. Three human LC3 isoforms undergo post-translational modifications during autophagy. Macroautophagy is the major inducible pathway for the general turnover of cytoplasmic constituents in eukaryotic cells, it is also responsible for the degradation of active cytoplasmic enzymes and organelles during nutrient starvation. Macroautophagy involves the formation of double-membrane bound autophagosomes which enclose the cytoplasmic constituent targeted for degradation in a membrane bound structure, which then fuse with the lysosome (or vacuole) releasing a single-membrane bound autophagic bodies which are then degraded within the lysosome (or vacuole). LC3B is a microtubule-associated protein that mediate the physical interactions between microtubules and components of the cytoskeleton. LC3B may play a role in processes involving cancer, aging, metabolic and neurodegenerative disorders and cardiovascular/pulmonary diseases.
Specifications
Specifications
LC3B
Polyclonal
DyLight™ 550
MAP1LC3B
Liquid
MAP1LC3B
A synthetic peptide made to an N-terminal portion of the human LC3 protein sequence (between residues 1-100).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bacteria, Bovine, Canine, Hamster, Human, Mouse, Non-human primate, Porcine, Rat, Zebrafish
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.75 mg/mL
50mM sodium borate with 0.05% sodium azide
O41515, Q9GZQ8, Q9CQV6, Q62625
RP11-346K17.1, ATG8E, LC3, LC3A, MAP1ALC3, MAP1BLC3, MAP1 light chain 3-like protein 1, MAP1A/1B light chain 3 A, MAP1A/MAP1B LC3 A, MAP1A/MAP1B light chain 3 A, autophagy-related protein LC3 A, autophagy-related ubiquitin-like modifier LC3 A, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A
Rabbit
IgG
100 μL
4° C, store in dark
Primary
100462754, 100769810, 322425, 408001, 479619, 64862, 67443, 81631