Learn More
anti-LC3/MAP1LC3A, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233254
Catalog No. NBP233254
Add to cart
Description
Description
LC3A Polyclonal antibody specifically detects LC3A in Human, Mouse, Rat, Bovine, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
Specifications
LC3A
0.5 mg/mL
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml
Q9H492
ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, lc3-i, ii, MAP1A/1B light chain 3 A, MAP1ALC3MAP1A/MAP1B LC3 A, MAP1BLC3MAP1A/MAP1B light chain 3 A, microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A
Rabbit
IgG
0.1 mg
Autophagy, Cancer, Cellular Markers, Cytoskeleton Markers, Hypoxia, Neuroscience, Phospho Specific, Signal Transduction
Primary
84557
Human, Mouse, Rat, Bovine, Opossum, Primate
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
0.2 ul PBS. BSA free - BSA free with No Preservative
Purified
MAP1LC3A
A portion of amino acids 1-50 of human ATG8 (LC3, MAP1LC3A, APG8) was used as the immunogen.
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
The amino acid sequence used as immunogen is 100% homologous in human, dog and mouse, 93% homologous in cow and Xenopus, 80% homologous in opossum and 73% homologous in pufferfish.