Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCAT Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526867

Catalog No. PIPA526867


Add to cart

Description

Description

LCAT encodes the extracellular cholesterol esterifying enzyme, lecithin-cholesterol acyltransferase. The esterification of cholesterol is required for cholesterol transport.
Specifications

Specifications

LCAT
Polyclonal
PBS with 0.09% sodium azide
P04180
phosphatidylcholine-sterol acyltransferase, phospholipid-cholesterol acyltransferase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3931
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LCAT
Liquid
LCAT
KLH conjugated synthetic peptide between 285-313 amino acids from the central region of human LCAT
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit