Learn More
Lck Mouse, Unlabeled, Clone: 28, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610097
Description
The p56[lck] protein kinase is a member of the src family of cytoplasmic protein-tyrosine kinases (PTKs). Members of this family have several common features: 1) unique N-terminal domains, 2) attachment to cellular membranes through a myristylated N-terminus, and 3) homologous SH2, SH3, and catalytic domains. Within the src family of PTKs, lck, fyn, and Yes are expressed in T cells. The unique N-terminal domain of p56[lck] interacts with the cytoplasmic tails of the CD4 and CD8 cell surface glycoproteins. CD4 and CD8 bind to surface major histocompatibility complex (MHC) class II and class I molecules, respectively. These complexes interact with the T cell antigen receptor (TCR) in the early stages of T cell activation. In addition, an activated lck kinase increases responsiveness of some T cell hybridomas to antigen. The phosphorylation status and, therefore, the activity of p56[lck] kinase is regulated by the CD45 tyrosine protein phosphatase. Several studies suggest that lck has many functions critical to T cell development and activation. Mice lacking a functional lck gene are drastically impaired in the production of T lymphocytes. Variants of the human Jurkat T cell line that do not express p56[lck] exhibit a diminished response to stimulation of the T cell receptor. Evidence suggests that lck is directly upstream from PI3-kinase in the signal transduction cascade in T cell activation.
Specifications
Lck | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human Lck aa. 1-191 | |
50 μg | |
Cell Biology | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
28 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.