Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-LD78 beta, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP142775

Catalog No. NBP142775


Add to cart

Description

Description

CCL3L1/LD78 beta Polyclonal antibody specifically detects CCL3L1/LD78 beta in Porcine, Bovine samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

CCL3L1/LD78 beta
LYOPH
Western Blot 1 ug/ml, ELISA 1:100-1:2000
Purified
CCL3L1
This protein-A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein raised in yeast, corresponding to the 70 amino acids of the mature porcine CCL3L1 protein.
Protein A purified
RUO
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is specific for swine CCL3L1 protein. A BLAST analysis was used to suggest cross-reactivity with CCL3L1 from bovine sources based on 100% identity with the immunizing sequence. Partial reactivity is expected against CCL3L1 from cat, dog, human, chimpanzee, baboon, bovine, and rat based on greater than 90% sequence homology. Cross-reactivity with CCL3L1 from other sources has not been determined.
Bovine, Porcine
ELISA, Western Blot
Unconjugated
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
C-C motif chemokine 3-like 1, CCL3L3, chemokine (C-C motif) ligand 3-like 1, D17S1718, G0/G1 switch regulatory protein 19-2, G0S19-2LD78, LD78BETA, LD78-beta(1-70), MGC104178, MGC12815, MGC182017, MIP1AP, PAT 464.2, SCYA3L, SCYA3L1464.2, small inducible cytokine A3-like 1, Small-inducible cytokine A3-like 1, Tonsillar lymphocyte LD78 beta protein
Rabbit
IgG
0.1 mg
Cell Cycle and Replication, Cytokine Research
Primary
6349
Reconstitute with 100 ul deionized water.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only