Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

lactate dehydrogenase A-like 6A, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human LDHAL6A protein.

Manufacturer:  Abnova Corporation H00160287B01

Catalog No. 89-008-632


Add to cart

Description

Description

Sequence: MATIKSELIKNFAEEEAIHHNKISIVGTGSVGVACAISILLKGLSDELVLVDVDEGKLKGETMDLQHGSPFMKMPNIVSSKDYLVTANSNLVIITAGARQKKGETRLDLVQRNVSIFKLMIPNITQYSPHCKLLIVTNPVDILTYVAWKLSGFPKNRVIGSGCNLDSARFRYFIGQRLGIHSESCHGLILGEHGDSSVPVWSGVNIAGVPLKDLNPDIGTDKDPEQWENVHKKK
Specifications

Specifications

lactate dehydrogenase A-like 6A
Unconjugated
No additive
BC014340.1
MGC23940
Mouse
50 uL
Endocrine/Metabolism
Y
Polyclonal
Human, Rat
Western Blot
Mouse polyclonal antibody raised against a full-length human LDHAL6A protein.
LDHAL6A
Serum
LDHAL6A
LDHAL6A (AAH14340.1, 1 a.a. ~ 234 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
160287
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit