Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDHD Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525637
Description
LDHD belongs to the D-isomerspecific 2-hydroxyacid dehydrogenase family. The similar protein inyeast has both D-lactate and D-glycerate dehydrogenase activities.Alternative splicing occurs at this locus and two transcriptvariants encoding distinct isoforms have been identified.Specifications
LDHD | |
Polyclonal | |
LDHD | |
DLD, D-lactate dehydrogenase, probable D-lactate dehydrogenase, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
197257 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q86WU2 | |
LDHD | |
KLH conjugated synthetic peptide between 35-62 amino acids from the N-terminal region of human LDHD | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction