Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDLR, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA131260

Catalog No. PA131260

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-31260 detects LDL from human samples.PA1-31260 has been successfully used in Western blot and ELISA applications.The PA1-31260 immunogen is full length plasma protein (Human).
Specifications

Specifications

LDL
Polyclonal
Unconjugated
LDLR
Purified
Chicken
IgY
50μg
Store at 4°C short term. For long term storage, Store at -20°C, avoiding freeze/thaw cycles.
Primary
3949
ELISA, Western Blot
1mg/mL
PBS with 0.02% sodium azide
P01130
LDLR
Full length plasma protein (Human).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit